Learn More
Medchemexpress LLC Naproxen (Standard) | 22204-53-1 | C14H14O3 | 100 MG
SDP
Catalog No. 5000375490
Encompass_Preferred
Naproxen (Standard) is an analytical and reference standard for research and analytical applications. This compound functions as a COX-1 and COX-2 inhibitor, with IC50s of 8.72 and 5.15 μM, respectively, in cell assays. It is commonly used in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • High purity of 99.91%
  • Inhibits both COX-1 and COX-2 enzymes
  • Suitable as an analytical and reference standard
Catalog No. 50-003-75490 Supplier Medchemexpress LLC Supplier No. HY15030R100MG
Naproxen (Standard) is an analytical and reference standard for research and analytical applications. This compound functions as a COX-1 and COX-2 inhibitor, with IC50s of 8.72 and 5.15 μM, respectively, in cell assays. It is commonly used in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • High purity of 99.91%
  • Inhibits both COX-1 and COX-2 enzymes
  • Suitable as an analytical and reference standard