Learn More
Medchemexpress LLC Naproxen (Standard) | 22204-53-1 | C14H14O3 | 250 MG
SDP
Catalog No. 5000374589
Encompass_Preferred
Naproxen (Standard) is the analytical standard of Naproxen, intended for research and analytical applications. It acts as a COX-1 and COX-2 inhibitor and is commonly used in qualitative, quantitative, and methodological research experiments. This product is for research use only.
  • Analytical standard
  • Intended for research and analytical applications
  • Functions as a COX-1 and COX-2 inhibitor
  • Used in HPLC, GC, and MS research experiments
  • IC50: 8.72 μM (COX-1)
  • IC50: 5.15 μM (COX-2)
Catalog No. 50-003-74589 Supplier Medchemexpress LLC Supplier No. 250MGHY15030R
Naproxen (Standard) is the analytical standard of Naproxen, intended for research and analytical applications. It acts as a COX-1 and COX-2 inhibitor and is commonly used in qualitative, quantitative, and methodological research experiments. This product is for research use only.
  • Analytical standard
  • Intended for research and analytical applications
  • Functions as a COX-1 and COX-2 inhibitor
  • Used in HPLC, GC, and MS research experiments
  • IC50: 8.72 μM (COX-1)
  • IC50: 5.15 μM (COX-2)