Learn More
Medchemexpress LLC Naproxen (Standard) | 22204-53-1 | C14H14O3 | 50 MG
SDP
Catalog No. 5000375378
Encompass_Preferred
Naproxen (Standard) is an analytical standard of Naproxen, designed for research and analytical applications. It acts as a COX-1 and COX-2 inhibitor with IC50s of 8.72 μM and 5.15 μM respectively in cell assays. This compound serves as a reference standard for assays and is frequently used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Acts as a COX-1 and COX-2 inhibitor
  • Serves as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-003-75378 Supplier Medchemexpress LLC Supplier No. HY15030R50MG
Naproxen (Standard) is an analytical standard of Naproxen, designed for research and analytical applications. It acts as a COX-1 and COX-2 inhibitor with IC50s of 8.72 μM and 5.15 μM respectively in cell assays. This compound serves as a reference standard for assays and is frequently used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Acts as a COX-1 and COX-2 inhibitor
  • Serves as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year