Learn More
Cayman Chemical (+)-Naproxen (sodium salt), Purity: ≥95%, CAS: 26159-34-2, 25 g
SDP
Catalog No. 502918155
Encompass_Preferred
An NSAID and COX inhibitor; inhibits COX-1 and COX-2 in COS-1 cells expressing the human enzymes (IC50s = 4.8 and 28.4 µM, respectively); inhibits TXB2 and PGE2 production ex vivo in isolated rat blood (IC50s = 5 and 13 µM, respectively)
Catalog No. 50-291-8155 Supplier Cayman Chemical Supplier No. 3755425 G
An NSAID and COX inhibitor; inhibits COX-1 and COX-2 in COS-1 cells expressing the human enzymes (IC50s = 4.8 and 28.4 µM, respectively); inhibits TXB2 and PGE2 production ex vivo in isolated rat blood (IC50s = 5 and 13 µM, respectively)