Learn More
AdipoGen NAPRTase B Subtilis rec
SDP
Catalog No. 502562763
Encompass_Preferred
Protein. Full-length NAPRTase Bacillus subtilis. Source E. coli. Endotoxin content N/A. Liquid. in 20mM TRIS-HCl, pH 7.8 containing 50mM sodium chloride. Purity >98% SDS-PAGE. Nicotinate Phosphoribosyltransferase NAPRTase is involved in the biological processes of pyridine nucleotide biosynthesis and nicotinate nucleotide salvage and functions by catalyzing the conversion of nicotinic acid NA to NA mononucleotide NaMN. It is a crucial factor in the NAD biosynthesis pathway. Catalytic activity Beta-nicotinate D-ribonucleotide diphosphate = nicotinate 5-phospho-alpha-D-ribose 1-diphosphate.
Catalog No. 50-256-2763 Supplier AdipoGen Supplier No. AG40T0104C050
Protein. Full-length NAPRTase Bacillus subtilis. Source E. coli. Endotoxin content N/A. Liquid. in 20mM TRIS-HCl, pH 7.8 containing 50mM sodium chloride. Purity >98% SDS-PAGE. Nicotinate Phosphoribosyltransferase NAPRTase is involved in the biological processes of pyridine nucleotide biosynthesis and nicotinate nucleotide salvage and functions by catalyzing the conversion of nicotinic acid NA to NA mononucleotide NaMN. It is a crucial factor in the NAD biosynthesis pathway. Catalytic activity Beta-nicotinate D-ribonucleotide diphosphate = nicotinate 5-phospho-alpha-D-ribose 1-diphosphate.