Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 9476MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: NAPSA/1239. Clonality: Monoclonal. Host: Mouse. Isotype: IgG kappa. Positive Control: Lung adenocarcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment of human Napsin-A protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 37kDa.Chromosome: 19q13.33. Gene Symbol: NAPSA. Gene ID: 9476. Ensemble: ENSG00000131400. Uniprot: XP_016883001.
Catalog No. 50-188-5346