Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 9476MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: NAPSA/1238. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Lung adenocarcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Napsin-A protein fragment (aa189-299). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 37kDa.Chromosome: 19q13.33. Gene Symbol: NAPSA. Gene ID: 9476. Ensemble: ENSG00000131400. Uniprot: XP_016883001.
Catalog No. 50-188-5345