Learn More
Sigma Aldrich Fine Chemicals Biosciences NAPTM-10 20 ST Cytiva 17-0
Catalog No. 502904887
Encompass
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.
Catalog No. 50-290-4887 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17085401
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.