Learn More
Sigma Aldrich Fine Chemicals Biosciences NAPTM 10 50 ST Cytiva 17 0
Catalog No. 502904727
Encompass
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.
Catalog No. 50-290-4727 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17085402
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.