Learn More
Sigma Aldrich Fine Chemicals Biosciences NAPTM 25 Cytiva 17 0852 02
Catalog No. 502907662
Encompass
NAP Columns are disposable columns prepacked with Sephadex G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.
Catalog No. 50-290-7662 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17085202
NAP Columns are disposable columns prepacked with Sephadex G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.