Learn More
Sigma Aldrich Fine Chemicals Biosciences NAPTM-5 20 ST Cytiva 17-08
Catalog No. 502901736
Encompass
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.
Catalog No. 50-290-1736 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17085301
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.