Learn More
Selleck Chemical LLC Naquotinib
SDP
Catalog No. 5000001487
Encompass_Preferred
Naquotinib (ASP8273) is an orally available irreversible mutant-selective epidermal growth factor receptor (EGFR) inhibitor with potential antineoplastic activity
Catalog No. 50-000-01487 Supplier Selleck Chemical LLC Supplier No. S84125MG
Naquotinib (ASP8273) is an orally available irreversible mutant-selective epidermal growth factor receptor (EGFR) inhibitor with potential antineoplastic activity