Learn More
Medchemexpress LLC Naquotinib mesylate | 1448237-05-5 | 98.1% | 658.81 | 1 ML
SDP
Catalog No. 5000401718
Encompass_Preferred
Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective, and irreversible EGFR inhibitor. It demonstrates potent inhibitory activity against EGFR mutants, with IC50 values ranging from 8 to 33 nM, while showing an IC50 of 230 nM for wild-type EGFR. This compound is primarily used in research contexts involving EGFR inhibition.
  • Orally available
  • Mutant-selective
  • Irreversible EGFR inhibitor
Catalog No. 50-004-01718 Supplier Medchemexpress LLC Supplier No. HY1980310MM/1ML
Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective, and irreversible EGFR inhibitor. It demonstrates potent inhibitory activity against EGFR mutants, with IC50 values ranging from 8 to 33 nM, while showing an IC50 of 230 nM for wild-type EGFR. This compound is primarily used in research contexts involving EGFR inhibition.
  • Orally available
  • Mutant-selective
  • Irreversible EGFR inhibitor