Learn More
Medchemexpress LLC Naquotinib mesylate | 1448237-05-5 | 98.1% | 658.81 | 25 MG
SDP
Catalog No. 5000403045
Encompass_Preferred
Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective, and irreversible EGFR inhibitor that demonstrates inhibitory concentrations (IC50s) ranging from 8 to 33 nM against EGFR mutants and 230 nM for wild-type EGFR. It effectively inhibits the growth of various EGFR-dependent non-small cell lung cancer cell lines, including those with exon 19 deletion and T790M resistance mutations. The compound selectively targets the phosphorylation of EGFR and its downstream signaling pathways, such as ERK and Akt, at concentrations as low as 10 nM in sensitive cell lines. Additionally, oral administration of this compound induces dose-dependent tumor regression in several xenograft models.
  • Orally available
  • Mutant-selective EGFR inhibitor
  • Irreversible EGFR inhibitor
  • Inhibits phosphorylation of EGFR and downstream signal pathway, ERK and Akt
  • Induces tumor shrinkage in EGFR mutated tumor models
Catalog No. 50-004-03045 Supplier Medchemexpress LLC Supplier No. 25MGHY19803
Naquotinib mesylate (ASP8273 mesylate) is an orally available, mutant-selective, and irreversible EGFR inhibitor that demonstrates inhibitory concentrations (IC50s) ranging from 8 to 33 nM against EGFR mutants and 230 nM for wild-type EGFR. It effectively inhibits the growth of various EGFR-dependent non-small cell lung cancer cell lines, including those with exon 19 deletion and T790M resistance mutations. The compound selectively targets the phosphorylation of EGFR and its downstream signaling pathways, such as ERK and Akt, at concentrations as low as 10 nM in sensitive cell lines. Additionally, oral administration of this compound induces dose-dependent tumor regression in several xenograft models.
  • Orally available
  • Mutant-selective EGFR inhibitor
  • Irreversible EGFR inhibitor
  • Inhibits phosphorylation of EGFR and downstream signal pathway, ERK and Akt
  • Induces tumor shrinkage in EGFR mutated tumor models