Learn More
Medchemexpress LLC Naquotinib mesylate | 1448237-05-5 | MFCD30496702 | 98.2% | 658.81 | C31H46N8O6S | 10 MG
SDP
Catalog No. 5000297489
Encompass_Preferred
Naquotinib mesylate is the mesylate salt of naquotinib, an orally available, irreversible, mutant-selective epidermal growth factor receptor (EGFR) inhibitor used in preclinical research. It preferentially inhibits common activating and resistance EGFR mutants while showing reduced potency against wild-type EGFR, making it useful for studies of mutant-selective signaling and resistance mechanisms.
  • Mutant-selective irreversible EGFR inhibitor
  • Potent against L858R, exon 19 deletion, and T790M mutants (IC50 8-33 nM)
  • Lower potency toward wild-type EGFR (IC50 ~230 nM)
  • Provided as a mesylate salt suitable for research applications
  • Molecular formula C31H46N8O6S; molecular weight 658.81
  • Reported research-grade purity approximately 98%
Catalog No. 50-002-97489 Supplier Medchemexpress LLC Supplier No. HY1980310MG
Naquotinib mesylate is the mesylate salt of naquotinib, an orally available, irreversible, mutant-selective epidermal growth factor receptor (EGFR) inhibitor used in preclinical research. It preferentially inhibits common activating and resistance EGFR mutants while showing reduced potency against wild-type EGFR, making it useful for studies of mutant-selective signaling and resistance mechanisms.
  • Mutant-selective irreversible EGFR inhibitor
  • Potent against L858R, exon 19 deletion, and T790M mutants (IC50 8-33 nM)
  • Lower potency toward wild-type EGFR (IC50 ~230 nM)
  • Provided as a mesylate salt suitable for research applications
  • Molecular formula C31H46N8O6S; molecular weight 658.81
  • Reported research-grade purity approximately 98%