Learn More
Cayman Chemical NArachidonyl Maleimide
SDP
Catalog No. 502887516
Encompass_Preferred
A potent inhibitor of monoacylglycerol lipase (IC50 = 140 nM in rat cerebellar membranes)
Catalog No. 50-288-7516 Supplier Cayman Chemical Supplier No. 1000751750 MG
A potent inhibitor of monoacylglycerol lipase (IC50 = 140 nM in rat cerebellar membranes)