Learn More
Medchemexpress LLC Naratriptan-d3 hydrochloride | 1190021-64-7 | 1 MG
SDP
Catalog No. 5000406175
Encompass_Preferred
Naratriptan-d3 is a deuterium labeled form of Naratriptan hydrochloride, a selective 5-HT1B/1D receptor agonist. This compound is peripherally active and exhibits good oral bioavailability, inducing cranial artery vasoconstriction. It also inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation and reduces sterile inflammation, making it suitable for research into acute migraine, targeting cranial vascular and neuroinflammatory mechanisms.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a selective 5-HT1B/1D receptor agonist
  • Exhibits good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
Catalog No. 50-004-06175 Supplier Medchemexpress LLC Supplier No. HYB0197AS1MG
Naratriptan-d3 is a deuterium labeled form of Naratriptan hydrochloride, a selective 5-HT1B/1D receptor agonist. This compound is peripherally active and exhibits good oral bioavailability, inducing cranial artery vasoconstriction. It also inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation and reduces sterile inflammation, making it suitable for research into acute migraine, targeting cranial vascular and neuroinflammatory mechanisms.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a selective 5-HT1B/1D receptor agonist
  • Exhibits good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation