Learn More
Medchemexpress LLC Naratriptan hydrochloride | 143388-64-1 | 100 MG
SDP
Catalog No. 5000407303
Encompass_Preferred
Naratriptan hydrochloride is an analytical standard intended for research and analytical applications. It is a selective 5-HT1B/1D receptor agonist that is peripherally active and exhibits good oral bioavailability. It induces cranial artery vasoconstriction by activating 5-HT1B/1D receptors, inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation, and reduces sterile inflammation.
  • Analytical standard for research and analytical applications
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active with good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Used in research of acute migraine
  • Commonly used in qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-07303 Supplier Medchemexpress LLC Supplier No. HYB0197A100MG
Naratriptan hydrochloride is an analytical standard intended for research and analytical applications. It is a selective 5-HT1B/1D receptor agonist that is peripherally active and exhibits good oral bioavailability. It induces cranial artery vasoconstriction by activating 5-HT1B/1D receptors, inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation, and reduces sterile inflammation.
  • Analytical standard for research and analytical applications
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active with good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Used in research of acute migraine
  • Commonly used in qualitative, quantitative, and methodological research experiments