Learn More
Medchemexpress LLC Naratriptan (hydrochloride) | 143388-64-1 | 99.7% | 1 ML
SDP
Catalog No. 5000405763
Encompass_Preferred
Naratriptan hydrochloride is a selective 5-HT1B/1D receptor agonist known for its peripheral activity and good oral bioavailability. It induces cranial artery vasoconstriction and inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation, reducing sterile inflammation. This compound is primarily used in acute migraine research, focusing on cranial vascular and neuroinflammatory mechanisms.
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active with good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Targets human recombinant 5-HT1B and 5-HT1D receptors
  • Used in research for acute migraine
Catalog No. 50-004-05763 Supplier Medchemexpress LLC Supplier No. HYB0197A10MM/1ML
Naratriptan hydrochloride is a selective 5-HT1B/1D receptor agonist known for its peripheral activity and good oral bioavailability. It induces cranial artery vasoconstriction and inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation, reducing sterile inflammation. This compound is primarily used in acute migraine research, focusing on cranial vascular and neuroinflammatory mechanisms.
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active with good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Targets human recombinant 5-HT1B and 5-HT1D receptors
  • Used in research for acute migraine