Learn More
Medchemexpress LLC Naratriptan (hydrochloride) | 143388-64-1 | 99.7% | 25 MG
SDP
Catalog No. 5000450662
Encompass_Preferred
Naratriptan hydrochloride (GR-85548A) is a selective 5-HT1B/1D receptor agonist. It is peripherally active and has good oral bioavailability, inducing cranial artery vasoconstriction by activating 5-HT1B/1D receptors. It also inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation and reduces sterile inflammation. This compound is primarily used in research for acute migraine, targeting cranial vascular and neuroinflammatory mechanisms.
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active
  • Good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Primarily used in research for acute migraine
Catalog No. 50-004-50662 Supplier Medchemexpress LLC Supplier No. 25MGHYB0197A
Naratriptan hydrochloride (GR-85548A) is a selective 5-HT1B/1D receptor agonist. It is peripherally active and has good oral bioavailability, inducing cranial artery vasoconstriction by activating 5-HT1B/1D receptors. It also inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation and reduces sterile inflammation. This compound is primarily used in research for acute migraine, targeting cranial vascular and neuroinflammatory mechanisms.
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active
  • Good oral bioavailability
  • Induces cranial artery vasoconstriction
  • Inhibits trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Primarily used in research for acute migraine