Learn More
Medchemexpress LLC Naratriptan hydrochloride | 143388-64-1 | 99.7% | 5 MG
SDP
Catalog No. 5000407335
Encompass_Preferred
Naratriptan hydrochloride (GR-85548A) is a selective 5-HT1B/1D receptor agonist. It is peripherally active and exhibits good oral bioavailability, causing cranial artery vasoconstriction by activating 5-HT1B/1D receptors. It also suppresses trigeminal nerve-mediated dural neurogenic plasma extravasation and reduces sterile inflammation. It is primarily used in acute migraine research, targeting cranial vascular and neuroinflammatory mechanisms.
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active with good oral bioavailability
  • Causes cranial artery vasoconstriction
  • Suppresses trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Targets human recombinant 5-HT1B and 5-HT1D receptors
  • Inhibits plasma protein extravasation in rat dural neurogenic inflammation model
  • High oral bioavailability in rats and dogs
Catalog No. 50-004-07335 Supplier Medchemexpress LLC Supplier No. HYB0197A5MG
Naratriptan hydrochloride (GR-85548A) is a selective 5-HT1B/1D receptor agonist. It is peripherally active and exhibits good oral bioavailability, causing cranial artery vasoconstriction by activating 5-HT1B/1D receptors. It also suppresses trigeminal nerve-mediated dural neurogenic plasma extravasation and reduces sterile inflammation. It is primarily used in acute migraine research, targeting cranial vascular and neuroinflammatory mechanisms.
  • Selective 5-HT1B/1D receptor agonist
  • Peripherally active with good oral bioavailability
  • Causes cranial artery vasoconstriction
  • Suppresses trigeminal nerve-mediated dural neurogenic plasma extravasation
  • Reduces sterile inflammation
  • Targets human recombinant 5-HT1B and 5-HT1D receptors
  • Inhibits plasma protein extravasation in rat dural neurogenic inflammation model
  • High oral bioavailability in rats and dogs