Learn More
Medchemexpress LLC Naratriptan hydrochloride | 143388-64-1 | MFCD08141821 | 99.7% | 371.93 g·mol⁻¹ | C17H26ClN3O2S | 10 MG
SDP
Catalog No. 5000297202
Encompass_Preferred
Naratriptan hydrochloride is the hydrochloride salt of naratriptan, a selective 5-HT1B/1D receptor agonist used as a research reagent to study migraine-related vascular responses and neuroinflammation. It is peripherally active, has good oral bioavailability, and induces cranial artery vasoconstriction by activating 5-HT1B/1D receptors.
  • High purity (99.69%).
  • Research applications: migraine pharmacology and cranial vascular studies.
  • Single small pack sizes suitable for analytical and in vitro use (e.g., 10 MG).
  • Well-characterized chemical identifiers (CAS 143388-64-1, MFCD08141821).
Catalog No. 50-002-97202 Supplier Medchemexpress LLC Supplier No. HYB0197A10MG
Naratriptan hydrochloride is the hydrochloride salt of naratriptan, a selective 5-HT1B/1D receptor agonist used as a research reagent to study migraine-related vascular responses and neuroinflammation. It is peripherally active, has good oral bioavailability, and induces cranial artery vasoconstriction by activating 5-HT1B/1D receptors.
  • High purity (99.69%).
  • Research applications: migraine pharmacology and cranial vascular studies.
  • Single small pack sizes suitable for analytical and in vitro use (e.g., 10 MG).
  • Well-characterized chemical identifiers (CAS 143388-64-1, MFCD08141821).