Learn More
Medchemexpress LLC Narciclasine | 29477-83-6 | 307.26 | 1 MG
SDP
Catalog No. 502029204
Encompass_Preferred
Narciclasine is a plant growth modulator that modulates the Rho/Rho kinase/LIM kinase/cofilin signaling pathway. It greatly increases GTPase RhoA activity and induces actin stress fiber formation in a RhoA-dependent manner.
  • Plant growth modulator
  • Modulates the Rho/Rho kinase/LIM kinase/cofilin signaling pathway
  • Increases GTPase RhoA activity
  • Induces actin stress fiber formation
  • Activates Rho and stress fibers in glioblastoma multiforme cells
  • Significantly increases survival in glioblastoma multiforme-bearing mice
  • Effective at lower doses compared to temozolomide
Catalog No. 50-202-9204 Supplier Medchemexpress LLC Supplier No. HY165631MG
 Restricted Item
Add to Cart
Add to Cart
Narciclasine is a plant growth modulator that modulates the Rho/Rho kinase/LIM kinase/cofilin signaling pathway. It greatly increases GTPase RhoA activity and induces actin stress fiber formation in a RhoA-dependent manner.
  • Plant growth modulator
  • Modulates the Rho/Rho kinase/LIM kinase/cofilin signaling pathway
  • Increases GTPase RhoA activity
  • Induces actin stress fiber formation
  • Activates Rho and stress fibers in glioblastoma multiforme cells
  • Significantly increases survival in glioblastoma multiforme-bearing mice
  • Effective at lower doses compared to temozolomide