Learn More
Cayman Chemical NarcIsosIde 10mg
SDP
Catalog No. 502937886
Encompass_Preferred
A flavonoid glycoside with antioxidative and antitumor activities; scavenges 27.74, 33.41, and 63% of DPPH radicals in a cell-free assay at 1, 10, and 100 µM, respectively; inhibits TPA-induced EBV-EA activation in Raji cells; inhibits TPA-induced papilloma formation in mice at 85 nmol
Catalog No. 50-293-7886 Supplier Cayman Chemical Supplier No. 2775910 MG
A flavonoid glycoside with antioxidative and antitumor activities; scavenges 27.74, 33.41, and 63% of DPPH radicals in a cell-free assay at 1, 10, and 100 µM, respectively; inhibits TPA-induced EBV-EA activation in Raji cells; inhibits TPA-induced papilloma formation in mice at 85 nmol