Learn More
Medchemexpress LLC HY-N0649 5mg Medchemexpress, Narcissin CAS:604-80-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06495MG
Encompass_Preferred
Medchemexpress, HY-N0649 5mg Narcissin CAS:604-80-8 Narcissin (Narcissoside), a flavonol glycoside isolated from Morinda citrifolia, exhibits evident scavenging activity against both authentic ONOO- and SIN-1-derived ONOO- with IC50s of 3.5 and 9.6 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1370