Learn More
Medchemexpress LLC Narcissin (narcissoside) | 604-80-8 | MFCD00017734 | 99.8% | 624.54 | C28H32O16 | 10 MG
SDP
Catalog No. 5000296144
Encompass_Preferred
Narcissin (narcissoside) is a flavonol glycoside research reagent with antioxidant activity. It scavenges peroxynitrite and is used in studies of oxidative stress and inflammatory signaling. The compound is a high-purity solid with molecular weight 624.54 g/mol and is supplied in small research quantities.
  • High purity: 99.82%.
  • Scavenges peroxynitrite (ONOO-) with IC50 = 3.5 μM (authentic) and 9.6 μM (SIN-1-derived).
  • Suitable for oxidative stress and inflammatory signaling assays.
  • Supplied as a solid for laboratory research applications.
  • Store protected from light; recommended storage 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-96144 Supplier Medchemexpress LLC Supplier No. HYN064910MG
Narcissin (narcissoside) is a flavonol glycoside research reagent with antioxidant activity. It scavenges peroxynitrite and is used in studies of oxidative stress and inflammatory signaling. The compound is a high-purity solid with molecular weight 624.54 g/mol and is supplied in small research quantities.
  • High purity: 99.82%.
  • Scavenges peroxynitrite (ONOO-) with IC50 = 3.5 μM (authentic) and 9.6 μM (SIN-1-derived).
  • Suitable for oxidative stress and inflammatory signaling assays.
  • Supplied as a solid for laboratory research applications.
  • Store protected from light; recommended storage 4°C; in solvent: -80°C (6 months), -20°C (1 month).