Learn More
Lakeside Manufacturing Inc Narcotic Cabinet Single Door, Double Lock, (2) Adjustable Shelves
Catalog No. 502884481
Encompass_Preferred
Perfect for areas short on space or for mobile transit of medications. The Lakeside LNC-2 Narcotic Cabinet has a single door with a double lock and (2) adjustable shelves. This model has a strong metal construction. The door can be installed to open from the left or right side. (2) Adjustable shelves to fit your needs. The mounting hardware is not included. Shipped through UPS/FedEx. Made in the USA.
Catalog No. 50-288-4481 Supplier Lakeside Manufacturing Inc Supplier No. LNC2
Perfect for areas short on space or for mobile transit of medications. The Lakeside LNC-2 Narcotic Cabinet has a single door with a double lock and (2) adjustable shelves. This model has a strong metal construction. The door can be installed to open from the left or right side. (2) Adjustable shelves to fit your needs. The mounting hardware is not included. Shipped through UPS/FedEx. Made in the USA.