Learn More
Omnimed Inc Single Door Narcotic Cabinet with 4 Shelves (24"H X 16"W X 8"D)
SDP
Catalog No. NC0762598
Encompass_Preferred
Omnimed narcotic cabinets are constructed of durable, heavy duty stainless steel and meet schedule 2 drug storage requirements. They have a strong piano hinged door and four adjustable aluminum shelves for added storage capacity. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right door access. The double wafer locks with heavy gauge locking cams keep contents secure. Three keys are included for each of the unique locks. Cabinets have pre-drilled holes that are reinforced with stainless steel mounting discs for wall attachment. Measuring 24H x 16W x 8D, these units are warranted for 10 years with 1 year on the locks.
Catalog No. NC0762598 Supplier Omnimed Inc Supplier No. 181481
Omnimed narcotic cabinets are constructed of durable, heavy duty stainless steel and meet schedule 2 drug storage requirements. They have a strong piano hinged door and four adjustable aluminum shelves for added storage capacity. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right door access. The double wafer locks with heavy gauge locking cams keep contents secure. Three keys are included for each of the unique locks. Cabinets have pre-drilled holes that are reinforced with stainless steel mounting discs for wall attachment. Measuring 24H x 16W x 8D, these units are warranted for 10 years with 1 year on the locks.