Learn More
Omnimed Inc Economy Double Door Narcotic Cabinet (15"H X 11"W X 8"D)
SDP
Catalog No. NC0116259
Encompass_Preferred
Cabinet is made of heavy gauge steel with a beige powder coated finish and are built to meet schedule 2 drug storage requirements. They have strong piano hinged doors and a two door, dual flat key locking system to ensure content security. The outside and inner door are secured with a heavy gauge locking cam. Three extra keys are included for each lock. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right outer door access. Its two aluminum shelves are adjustable and increase storage capacity. Pre-drilled holes ensure stable wall attachment. Warranted for LIFE with 1 year on the locks.
Catalog No. NC0116259 Supplier Omnimed Inc Supplier No. 182150
Cabinet is made of heavy gauge steel with a beige powder coated finish and are built to meet schedule 2 drug storage requirements. They have strong piano hinged doors and a two door, dual flat key locking system to ensure content security. The outside and inner door are secured with a heavy gauge locking cam. Three extra keys are included for each lock. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right outer door access. Its two aluminum shelves are adjustable and increase storage capacity. Pre-drilled holes ensure stable wall attachment. Warranted for LIFE with 1 year on the locks.