Learn More
Cayman Chemical Lycorine, Purity: ≥95%, CAS: 476-28-8, 50 mg
SDP
Catalog No. 502992151
Encompass_Preferred
An alkaloid with diverse biological activities; active against P. falciparum (IC50 = 1.026 µg/ml); inhibits SARS-CoV viral infection in Vero cells (EC50 = 15.7 µM); inhibits migration of A549 NSCLC and U373 MG glioblastoma cells at 5 µM; increases survival and reduces virus-induced paralysis in a mouse model of enterovirus 71 infection at 0.4 mg/kg
Catalog No. 50-299-2151 Supplier Cayman Chemical Supplier No. 3065050 MG
An alkaloid with diverse biological activities; active against P. falciparum (IC50 = 1.026 µg/ml); inhibits SARS-CoV viral infection in Vero cells (EC50 = 15.7 µM); inhibits migration of A549 NSCLC and U373 MG glioblastoma cells at 5 µM; increases survival and reduces virus-induced paralysis in a mouse model of enterovirus 71 infection at 0.4 mg/kg