Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nardosinone | 23720-80-1 | 250.33 | 5 MG
SDP

Catalog No. 5000389799
Encompass_Preferred

Nardosinone, isolated from Nardostachys chinensis, is identified as the first enhancer of the neuritogenic action of dbcAMP and staurosporine. It may serve as a pharmacological tool for studying the mechanism of action of nerve growth factor (NGF) and other neuritogenic substances.

  • Purity of 99.81%
  • Appearance as a white to off-white solid
  • Store at 4°C, protected from light; in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
  • Initial source from plants, Valerianaceae, Nardostachys jatamansi (D. Don) DC.
  • Classified as a terpenoid, sesquiterpene
  • Exhibits antiinflammatory activity in RAW264.7 cells with an IC50 of 11.1 μM

Catalog No. 50-003-89799 Supplier Medchemexpress LLC Supplier No. HYN03805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nardosinone, isolated from Nardostachys chinensis, is identified as the first enhancer of the neuritogenic action of dbcAMP and staurosporine. It may serve as a pharmacological tool for studying the mechanism of action of nerve growth factor (NGF) and other neuritogenic substances.

  • Purity of 99.81%
  • Appearance as a white to off-white solid
  • Store at 4°C, protected from light; in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
  • Initial source from plants, Valerianaceae, Nardostachys jatamansi (D. Don) DC.
  • Classified as a terpenoid, sesquiterpene
  • Exhibits antiinflammatory activity in RAW264.7 cells with an IC50 of 11.1 μM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.