Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nardosinone | 23720-80-1 | 99.5% | 250.33 | C15H22O3 | 10 MG
SDP

Catalog No. 5000296486
Encompass_Preferred

Nardosinone is a naturally derived small molecule reported to enhance neuritogenic responses to dbcAMP and staurosporine. It is supplied as a high-purity research reagent with documented solubility and storage guidance for experimental use.

  • High purity (~99.5%).
  • Molecular formula C15H22O3; molecular weight 250.33 g/mol.
  • Available as a solid, white to off-white; common package size 10 MG.
  • Soluble in DMSO (100 mg/mL); in vivo formulations noted with 10% DMSO plus SBE-β-CD or corn oil.
  • Store at 4°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-96486 Supplier Medchemexpress LLC Supplier No. HYN038010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nardosinone is a naturally derived small molecule reported to enhance neuritogenic responses to dbcAMP and staurosporine. It is supplied as a high-purity research reagent with documented solubility and storage guidance for experimental use.

  • High purity (~99.5%).
  • Molecular formula C15H22O3; molecular weight 250.33 g/mol.
  • Available as a solid, white to off-white; common package size 10 MG.
  • Soluble in DMSO (100 mg/mL); in vivo formulations noted with 10% DMSO plus SBE-β-CD or corn oil.
  • Store at 4°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.