Learn More
Aviva Systems Biology Corp NARG1 Antibody - middle region (ARP48990_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NARG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NARG1. Alternate names: Ga19, NATH, TBDN100, NARG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59987