Learn More
AdipoGen Nargenicin A1
SDP
Supplier:  AdipoGen BVT0204M005
Encompass_Preferred
Chemical. CAS 70695-02-2. Formula C28H37NO8. MW 515.6. Isolated from Actinomyces sp. Gö301. Antibiotic against Gram-positive bacteria, particularly Staphylococcus and Clostridia. More effective against multi-resistant strains MRSA than erythromycin and vancomycin. Inhibits cell proliferation and induces HL-60 cell differentiation in combination with 1,25-Dihydroxyvitamin-D3 and ATRA. Displays low cytotoxicity compared to erythromycin and spiramycin.
Catalog No. 50-256-2063