Learn More
Abcam Nargenicin A1, Antibiotic agent, 5MG
Catalog No. 502705894
Encompass_Preferred
MW 515.6 Da, Purity >97%. Broad spectrum antibiotic agent. Potential anti-inflammatory agent. Active against Gram-positive bacteria, particularly Staphylococcus and Clostridia. Attenuates LPS-induced nitric oxide production in microglia. Suppresses IL-1β, TNF-α and iNOS production.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5894 Supplier Abcam Supplier No. AB1443125MG
MW 515.6 Da, Purity >97%. Broad spectrum antibiotic agent. Potential anti-inflammatory agent. Active against Gram-positive bacteria, particularly Staphylococcus and Clostridia. Attenuates LPS-induced nitric oxide production in microglia. Suppresses IL-1β, TNF-α and iNOS production.
The product is subject to the following: Abcam Restricted Use Statement