Learn More
TARGETMOL CHEMICALS INC Naringenin 1G
SDP
Catalog No. 502879885
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Naringenin (NSC-11855) is a flavanone that is considered to have a bioactive effect on human health as antioxidant, free radical scavenger, antiinflammatory, carbohydrate metabolism promoter, immunity system modulater. This substance has also been shown to repair DNA. Purity 98.81%
Catalog No. 50-287-9885 Supplier TARGETMOL CHEMICALS INC Supplier No. T28381G
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Naringenin (NSC-11855) is a flavanone that is considered to have a bioactive effect on human health as antioxidant, free radical scavenger, antiinflammatory, carbohydrate metabolism promoter, immunity system modulater. This substance has also been shown to repair DNA. Purity 98.81%