Learn More
Selleck Chemical LLC Naringenin S2394-25mg
SDP
Catalog No. 503112933
Encompass_Preferred
Naringenin (NSC 34875 S-Dihydrogenistein NSC 11855 Salipurol) is a natural predominant flavanone derived from plant food which is considered to have a bioactive effect on human health as antioxidant free radical scavenger anti-inflammatory carbohydrate metabolism promoter and immune system modulator
Catalog No. 50-311-2933 Supplier Selleck Chemical LLC Supplier No. S239425MG
Naringenin (NSC 34875 S-Dihydrogenistein NSC 11855 Salipurol) is a natural predominant flavanone derived from plant food which is considered to have a bioactive effect on human health as antioxidant free radical scavenger anti-inflammatory carbohydrate metabolism promoter and immune system modulator