Learn More
Apexbio Technology LLC Naringenin 480-41-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094338
Encompass_Preferred
Naringenin (CAS 480-41-1) is a small-molecule inhibitor designed to modulate inflammatory and antiviral pathways thereby regulating cellular inflammatory responses and viral replication Naringenin exerts its biological activity primarily through inhibition of the NF- B pathway resulting in reduced inflammatory cytokine production Additionally it demonstrates antioxidative properties by scavenging reactive oxygen species (ROS) In in vitro studies naringenin exhibits inhibition of dengue virus replication with reported IC50 values ranging from approximately 52 64 M to 143 M depending on experimental conditions and viral strains Based on these pharmacological properties naringenin holds research potential in inflammation oxidative stress-related cellular models and dengue virus infection studies
Catalog No. 50-000-94338 Supplier Apexbio Technology LLC Supplier No. N13705.1
Naringenin (CAS 480-41-1) is a small-molecule inhibitor designed to modulate inflammatory and antiviral pathways thereby regulating cellular inflammatory responses and viral replication Naringenin exerts its biological activity primarily through inhibition of the NF- B pathway resulting in reduced inflammatory cytokine production Additionally it demonstrates antioxidative properties by scavenging reactive oxygen species (ROS) In in vitro studies naringenin exhibits inhibition of dengue virus replication with reported IC50 values ranging from approximately 52 64 M to 143 M depending on experimental conditions and viral strains Based on these pharmacological properties naringenin holds research potential in inflammation oxidative stress-related cellular models and dengue virus infection studies