Learn More
Medchemexpress LLC Naringenin | 480-41-1 | 99.28% | 272.25 | 25 G
SDP
Catalog No. 5000452673
Encompass_Preferred
Naringenin is a predominant flavanone that exhibits strong anti-inflammatory and antioxidant activities. It also possesses anti-dengue virus (DENV) activity.
  • Inhibits proliferation and induces apoptosis in HepG2 cells.
  • Causes cell cycle arrest and increased caspase-3 activity in A431 cells.
  • Reduces triglycerides and cholesterol in plasma and liver.
  • Lowers adiposity and increases PPARα protein expression in the liver.
Catalog No. 50-004-52673 Supplier Medchemexpress LLC Supplier No. 25GHYW011641
Naringenin is a predominant flavanone that exhibits strong anti-inflammatory and antioxidant activities. It also possesses anti-dengue virus (DENV) activity.
  • Inhibits proliferation and induces apoptosis in HepG2 cells.
  • Causes cell cycle arrest and increased caspase-3 activity in A431 cells.
  • Reduces triglycerides and cholesterol in plasma and liver.
  • Lowers adiposity and increases PPARα protein expression in the liver.