Learn More
Medchemexpress LLC Naringenin | 480-41-1 | 99.3% | 272.25 | 5 MG
SDP
Catalog No. 5000427505
Encompass_Preferred
Naringenin is the predominant flavanone found in Citrus reticulata Blanco. It exhibits strong anti-inflammatory and antioxidant activities, and also possesses anti-dengue virus (DENV) activity.
  • Inhibits proliferation and induces apoptosis in HepG2 cells
  • Reduces cell viability and increases DNA fragmentation in A431 cells
  • Reduces plasma and liver triglyceride and cholesterol levels
  • Lowers adiposity and triglyceride content
  • Increases hepatic PPARα protein expression and enhances CPT-1 and UCP2 expression
  • Boosts hepatic fatty acid oxidation and prevents sterol regulatory element-binding protein 1c-mediated lipogenesis
  • Inhibits TNF-α-induced VSMC proliferation and migration
Catalog No. 50-004-27505 Supplier Medchemexpress LLC Supplier No. HYN01005MG
Naringenin is the predominant flavanone found in Citrus reticulata Blanco. It exhibits strong anti-inflammatory and antioxidant activities, and also possesses anti-dengue virus (DENV) activity.
  • Inhibits proliferation and induces apoptosis in HepG2 cells
  • Reduces cell viability and increases DNA fragmentation in A431 cells
  • Reduces plasma and liver triglyceride and cholesterol levels
  • Lowers adiposity and triglyceride content
  • Increases hepatic PPARα protein expression and enhances CPT-1 and UCP2 expression
  • Boosts hepatic fatty acid oxidation and prevents sterol regulatory element-binding protein 1c-mediated lipogenesis
  • Inhibits TNF-α-induced VSMC proliferation and migration