Learn More
Indofine Chemical Naringenin, 98.2%, 5 Gm
SDP
Catalog No. 50908108
Encompass_Preferred
Moelcular Formula: C15H12O5, Purity: 98%, Molecular Weight: 272.26, Appearance: Brown crystalline powder, Solubility: Soluble in alcohol, ether, benzene, Storage conditions: Store in a cool, dry place, Also known as 5,7,4'-Trihydroxyflavanone; 2,3-dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
Catalog No. 50-908-108 Supplier Indofine Chemical Supplier No. N101 5 GM
Moelcular Formula: C15H12O5, Purity: 98%, Molecular Weight: 272.26, Appearance: Brown crystalline powder, Solubility: Soluble in alcohol, ether, benzene, Storage conditions: Store in a cool, dry place, Also known as 5,7,4'-Trihydroxyflavanone; 2,3-dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one