Learn More
Medchemexpress LLC (±)-Naringenin | 67604-48-2 | 99.8% | 272.26 | 50 G
SDP
Catalog No. 5000455082
Encompass_Preferred
(±)-Naringenin is an orally available anti-inflammatory agent that can regulate both acute and chronic inflammation responses, while also showing antioxidant, neuroprotective, liver-protective, and anti-cancer effects. It promotes vasodilation in endothelial cells by activating BKCa channels in muscle cells and exerts protective effects against experimental colitis by inhibiting Toll-like receptor 4/NF-κB signaling, making it useful in studies related to sepsis, fulminant hepatitis, fibrosis, and cancer research.
  • Regulates acute and chronic inflammation responses
  • Exhibits antioxidant effects
  • Provides neuroprotective benefits
  • Offers liver-protective properties
  • Shows anti-cancer effects
  • Promotes vasodilation in endothelial cells
  • Inhibits Toll-like receptor 4/NF-κB signaling
Catalog No. 50-004-55082 Supplier Medchemexpress LLC Supplier No. HYW01164150G
(±)-Naringenin is an orally available anti-inflammatory agent that can regulate both acute and chronic inflammation responses, while also showing antioxidant, neuroprotective, liver-protective, and anti-cancer effects. It promotes vasodilation in endothelial cells by activating BKCa channels in muscle cells and exerts protective effects against experimental colitis by inhibiting Toll-like receptor 4/NF-κB signaling, making it useful in studies related to sepsis, fulminant hepatitis, fibrosis, and cancer research.
  • Regulates acute and chronic inflammation responses
  • Exhibits antioxidant effects
  • Provides neuroprotective benefits
  • Offers liver-protective properties
  • Shows anti-cancer effects
  • Promotes vasodilation in endothelial cells
  • Inhibits Toll-like receptor 4/NF-κB signaling