Learn More
Medchemexpress LLC (±)-Naringenin | 67604-48-2 | MFCD00006844 | 99.83% | 272.26 | 1 ML
SDP
Catalog No. 5000454959
Encompass_Preferred
(±)-Naringenin is an orally available anti-inflammatory agent that regulates acute and chronic inflammation, and exhibits antioxidant, neuroprotective, liver-protective, and anti-cancer effects. It promotes vasodilation and protects against experimental colitis by inhibiting Toll-like receptor 4/NF-κB signaling, useful in studies of sepsis, fulminant hepatitis, fibrosis, and cancer. It appears as an off-white to light yellow solid.
  • Regulates acute and chronic inflammation.
  • Antioxidant, neuroprotective, liver-protective, and anti-cancer effects.
  • Promotes vasodilation.
  • Protects against experimental colitis.
  • Useful in sepsis, hepatitis, fibrosis, and cancer research.
  • Orally available.
Catalog No. 50-004-54959 Supplier Medchemexpress LLC Supplier No. HYW01164110MM/1ML
(±)-Naringenin is an orally available anti-inflammatory agent that regulates acute and chronic inflammation, and exhibits antioxidant, neuroprotective, liver-protective, and anti-cancer effects. It promotes vasodilation and protects against experimental colitis by inhibiting Toll-like receptor 4/NF-κB signaling, useful in studies of sepsis, fulminant hepatitis, fibrosis, and cancer. It appears as an off-white to light yellow solid.
  • Regulates acute and chronic inflammation.
  • Antioxidant, neuroprotective, liver-protective, and anti-cancer effects.
  • Promotes vasodilation.
  • Protects against experimental colitis.
  • Useful in sepsis, hepatitis, fibrosis, and cancer research.
  • Orally available.