Learn More
TARGETMOL CHEMICALS INC Naringenin chalcone 10MG
SDP
Catalog No. 502877014
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Naringenin chalcone is a natural product isolated from the stem of the plant Machaerium isadelphum (Fabaceae).Naringenin chalcone has anti-inflammatory activity. Purity 98.11%
Catalog No. 50-287-7014 Supplier TARGETMOL CHEMICALS INC Supplier No. T778410MG
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Naringenin chalcone is a natural product isolated from the stem of the plant Machaerium isadelphum (Fabaceae).Naringenin chalcone has anti-inflammatory activity. Purity 98.11%