Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naringenin chalcone | 73692-50-9 | 1 ML
SDP

Catalog No. 5000421751
Encompass_Preferred

Naringenin chalcone is an orally active intermediate involved in flavonol biosynthesis. It induces apoptosis, inhibits the production of MCP-1 and NO, and exhibits anticancer activity against glioblastoma. This compound also possesses anti-inflammatory and anti-allergic properties.

  • Induces apoptosis
  • Inhibits the production of MCP-1 and NO
  • Exhibits anticancer activity against glioblastoma
  • Possesses anti-inflammatory properties
  • Possesses anti-allergic properties
  • High purity: 98.86%
  • Molecular weight: 272.25
  • Formula: C15H12O5
  • Appearance: Solid, light yellow to orange

Catalog No. 50-004-21751 Supplier Medchemexpress LLC Supplier No. HYN300710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naringenin chalcone is an orally active intermediate involved in flavonol biosynthesis. It induces apoptosis, inhibits the production of MCP-1 and NO, and exhibits anticancer activity against glioblastoma. This compound also possesses anti-inflammatory and anti-allergic properties.

  • Induces apoptosis
  • Inhibits the production of MCP-1 and NO
  • Exhibits anticancer activity against glioblastoma
  • Possesses anti-inflammatory properties
  • Possesses anti-allergic properties
  • High purity: 98.86%
  • Molecular weight: 272.25
  • Formula: C15H12O5
  • Appearance: Solid, light yellow to orange

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.