Learn More
Medchemexpress LLC Naringenin chalcone | 73692-50-9 | 5 MG
SDP
Catalog No. 5000420978
Encompass_Preferred
Naringenin chalcone is an orally active intermediate involved in flavonol biosynthesis. It induces apoptosis, inhibits the production of MCP-1 and NO, and exhibits anticancer activity against glioblastoma. It also possesses anti-inflammatory and anti-allergic properties.
  • Intermediate in flavonol biosynthesis
  • Induces apoptosis
  • Inhibits production of MCP-1 and NO
  • Exhibits anticancer activity against glioblastoma
  • Possesses anti-inflammatory properties
  • Possesses anti-allergic properties
Catalog No. 50-004-20978 Supplier Medchemexpress LLC Supplier No. HYN30075MG
Naringenin chalcone is an orally active intermediate involved in flavonol biosynthesis. It induces apoptosis, inhibits the production of MCP-1 and NO, and exhibits anticancer activity against glioblastoma. It also possesses anti-inflammatory and anti-allergic properties.
  • Intermediate in flavonol biosynthesis
  • Induces apoptosis
  • Inhibits production of MCP-1 and NO
  • Exhibits anticancer activity against glioblastoma
  • Possesses anti-inflammatory properties
  • Possesses anti-allergic properties