Learn More
Medchemexpress LLC Naringenin chalcone | 73692-50-9 | MFCD00016762 | 100 MG
SDP
Catalog No. 5000421247
Encompass_Preferred
Naringenin chalcone is an orally active intermediate in flavonol biosynthesis. It induces apoptosis and inhibits the production of MCP-1 and NO. This compound exhibits anticancer activity against glioblastoma and possesses anti-inflammatory and anti-allergic properties. Intended for research use only.
  • Induces apoptosis in glioblastoma cells.
  • Inhibits production of MCP-1, TNF-α, and NO in macrophages.
  • Increases adiponectin mRNA levels and protein secretion in adipocytes.
  • Reduces eosinophilic airway inflammation and airway hyperresponsiveness.
  • Suppresses Th2 cytokine production and reduces mucus overproduction.
  • Reduces tumor volume and weight in glioblastoma xenograft models.
  • Increases plasma adiponectin levels in diabetic mice.
  • Inhibits anaphylactic response to IgE-mediated passive cutaneous anaphylaxis.
Catalog No. 50-004-21247 Supplier Medchemexpress LLC Supplier No. HYN3007100MG
Naringenin chalcone is an orally active intermediate in flavonol biosynthesis. It induces apoptosis and inhibits the production of MCP-1 and NO. This compound exhibits anticancer activity against glioblastoma and possesses anti-inflammatory and anti-allergic properties. Intended for research use only.
  • Induces apoptosis in glioblastoma cells.
  • Inhibits production of MCP-1, TNF-α, and NO in macrophages.
  • Increases adiponectin mRNA levels and protein secretion in adipocytes.
  • Reduces eosinophilic airway inflammation and airway hyperresponsiveness.
  • Suppresses Th2 cytokine production and reduces mucus overproduction.
  • Reduces tumor volume and weight in glioblastoma xenograft models.
  • Increases plasma adiponectin levels in diabetic mice.
  • Inhibits anaphylactic response to IgE-mediated passive cutaneous anaphylaxis.