Learn More
Medchemexpress LLC Naringenin chalcone | 73692-50-9 | MFCD00016762 | 25 MG
SDP
Catalog No. 5000421575
Encompass_Preferred
Naringenin chalcone is an orally active intermediate in flavonol biosynthesis. It functions as a metabolite and is known for its anti-allergic and anti-inflammatory properties. This chalcone is also noted for inducing apoptosis and inhibiting the production of MCP-1 and NO, as well as exhibiting anticancer activity against glioblastoma. This product is intended for research use only.
  • Intermediate in flavonol biosynthesis
  • Induces apoptosis
  • Inhibits the production of MCP-1 and NO
  • Exhibits anticancer activity against glioblastoma
  • Possesses anti-inflammatory properties
  • Possesses anti-allergic properties
  • Functions as a metabolite
  • Intended for research use only
Catalog No. 50-004-21575 Supplier Medchemexpress LLC Supplier No. 25MGHYN3007
Naringenin chalcone is an orally active intermediate in flavonol biosynthesis. It functions as a metabolite and is known for its anti-allergic and anti-inflammatory properties. This chalcone is also noted for inducing apoptosis and inhibiting the production of MCP-1 and NO, as well as exhibiting anticancer activity against glioblastoma. This product is intended for research use only.
  • Intermediate in flavonol biosynthesis
  • Induces apoptosis
  • Inhibits the production of MCP-1 and NO
  • Exhibits anticancer activity against glioblastoma
  • Possesses anti-inflammatory properties
  • Possesses anti-allergic properties
  • Functions as a metabolite
  • Intended for research use only