Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naringenin chalcone | 73692-50-9 | MFCD00016762 | 272.25 g/mol | C15H12O5 | 10 MG
SDP

Catalog No. 5000289163
Encompass_Preferred

Naringenin chalcone is a tetrahydroxychalcone flavonoid intermediate supplied for laboratory research and analytical applications. It is used in studies of flavonol biosynthesis and in biological assays where its reported activities (apoptosis induction, anti-inflammatory, anti-allergic, and anticancer effects) are relevant.

  • Research reagent for studies of flavonol biosynthesis.
  • Reported biological activities include apoptosis induction and anti-inflammatory effects.
  • Suitable for analytical applications and small-scale laboratory assays.
  • Molecular weight 272.25 g/mol; chemical formula C15H12O5.
  • CAS number 73692-50-9; supplied in small package sizes for research use.

Catalog No. 50-002-89163 Supplier Medchemexpress LLC Supplier No. HYN300710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naringenin chalcone is a tetrahydroxychalcone flavonoid intermediate supplied for laboratory research and analytical applications. It is used in studies of flavonol biosynthesis and in biological assays where its reported activities (apoptosis induction, anti-inflammatory, anti-allergic, and anticancer effects) are relevant.

  • Research reagent for studies of flavonol biosynthesis.
  • Reported biological activities include apoptosis induction and anti-inflammatory effects.
  • Suitable for analytical applications and small-scale laboratory assays.
  • Molecular weight 272.25 g/mol; chemical formula C15H12O5.
  • CAS number 73692-50-9; supplied in small package sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.