Learn More
Medchemexpress LLC Naringenin-d4 | 1192260-78-8 | MFCD34676368 | 99.9% | 276.28 | C15H8D4O5 | 10 MG
SDP
Catalog No. 5000296318
Encompass_Preferred
Naringenin-d4 is the deuterated (stable isotope-labelled) analogue of the flavanone naringenin, supplied as a solid reference standard for analytical, tracer, and metabolic studies. It is commonly used as an internal standard in mass spectrometry and for pharmacokinetic or mechanistic investigations.
  • Deuterium-labelled analogue suitable as an internal standard for LC-MS/MS.
  • High chemical purity for reliable quantitative analysis.
  • Supplied as a dry powder for convenient handling and storage.
  • Available in small package sizes, including 10 mg, for analytical workflows.
  • Structural identifiers provided for cheminformatics and method development.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-002-96318 Supplier Medchemexpress LLC Supplier No. HYN0100S10MG
Naringenin-d4 is the deuterated (stable isotope-labelled) analogue of the flavanone naringenin, supplied as a solid reference standard for analytical, tracer, and metabolic studies. It is commonly used as an internal standard in mass spectrometry and for pharmacokinetic or mechanistic investigations.
  • Deuterium-labelled analogue suitable as an internal standard for LC-MS/MS.
  • High chemical purity for reliable quantitative analysis.
  • Supplied as a dry powder for convenient handling and storage.
  • Available in small package sizes, including 10 mg, for analytical workflows.
  • Structural identifiers provided for cheminformatics and method development.
  • Stable when stored under recommended conditions for long-term use.